Learn More
Medchemexpress LLC Tetrazine-ph-acid | 1380500-92-4 | MFCD24503098 | 98.7% | 216.20 | C10H8N4O2 | 50MG
SDP
Catalog No. 5000236034
Encompass_Preferred
Tetrazine-Ph-acid is a tetrazine-containing carboxylic acid reagent used for rapid bioorthogonal inverse electron-demand Diels-Alder (iEDDA) reactions. It serves as a click-chemistry building block and linker component for PROTAC synthesis and conjugation workflows. The compound is supplied as a colored solid with high reported purity and is intended for research applications under recommended cold storage conditions.
  • Tetrazine functional group enabling fast iEDDA click reactions.
  • Suitable as a linker building block in PROTAC and conjugation chemistry.
  • High reported purity (98.7%) for synthetic applications.
  • Molecular weight 216.20 g/mol; formula C10H8N4O2.
  • Pink to red solid, soluble in DMSO and common organic solvents.
  • Stable when stored cold; follow supplier storage recommendations.
Catalog No. 50-002-36034 Supplier Medchemexpress LLC Supplier No. HY12448050MG
Tetrazine-Ph-acid is a tetrazine-containing carboxylic acid reagent used for rapid bioorthogonal inverse electron-demand Diels-Alder (iEDDA) reactions. It serves as a click-chemistry building block and linker component for PROTAC synthesis and conjugation workflows. The compound is supplied as a colored solid with high reported purity and is intended for research applications under recommended cold storage conditions.
  • Tetrazine functional group enabling fast iEDDA click reactions.
  • Suitable as a linker building block in PROTAC and conjugation chemistry.
  • High reported purity (98.7%) for synthetic applications.
  • Molecular weight 216.20 g/mol; formula C10H8N4O2.
  • Pink to red solid, soluble in DMSO and common organic solvents.
  • Stable when stored cold; follow supplier storage recommendations.