Learn More
Medchemexpress LLC Tetrazine-SS-NHS | 95.0% | 476.53 g/mol | C19H20N6O5S2 | 5 MG
SDP
Catalog No. 5000204507
Encompass_Preferred
Tetrazine-SS-NHS is a cleavable heterobifunctional linker and click chemistry reagent containing a tetrazine moiety and an NHS ester for bioconjugation. It is supplied as an orange-red solid and is used for inverse electron-demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) groups and for constructing cleavable antibody-drug conjugates.
  • Cleavable disulfide (SS) linker for controlled release.
  • Tetrazine moiety enables rapid iEDDA click reactions with TCO-functionalized partners.
  • NHS ester facilitates amine coupling to biomolecules.
  • Supplied as an orange-red solid with 95.0% purity.
  • Recommended storage: powder at -20°C; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-04507 Supplier Medchemexpress LLC Supplier No. HY1360325MG
Tetrazine-SS-NHS is a cleavable heterobifunctional linker and click chemistry reagent containing a tetrazine moiety and an NHS ester for bioconjugation. It is supplied as an orange-red solid and is used for inverse electron-demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) groups and for constructing cleavable antibody-drug conjugates.
  • Cleavable disulfide (SS) linker for controlled release.
  • Tetrazine moiety enables rapid iEDDA click reactions with TCO-functionalized partners.
  • NHS ester facilitates amine coupling to biomolecules.
  • Supplied as an orange-red solid with 95.0% purity.
  • Recommended storage: powder at -20°C; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).