Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™

Catalog No. AC432990010
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432990010 1 L AcroSeal™ Glass Bottle
AC432991000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432990010 Supplier Thermo Scientific Chemicals Supplier No. 432990010
Add to Cart

CAS: 288-94-8 | CH2N4 | 70.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrazole
CAS 288-94-8, 75-05-8
Color Cream to Orange
Density 0.8000g/mL
Boiling Point 77°C
Flash Point 5°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula CH2N4
Quantity 1 L
Specific Gravity 0.8
Synonym 1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
InChI Key KJUGUADJHNHALS-UHFFFAOYSA-N
SMILES C1=NNN=N1
IUPAC Name 2H-tetrazole
Molecular Weight (g/mol) 70.05
PubChem CID 67519
ChEBI CHEBI:33193
Formula Weight 70.05
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.