Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrazolium Violet 98.0+%, TCI America™
SDP

Catalog No. T01741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01741G 1 g
T017410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01741G Supplier TCI America Supplier No. T01741G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1719-71-7
Molecular Formula C23H17ClN4
Molecular Weight (g/mol) 384.867
MDL Number MFCD00011875
InChI Key RONADMZTCCPLEF-UHFFFAOYSA-M
Synonym tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at
PubChem CID 74395
ChEBI CHEBI:75193
IUPAC Name 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-]

Specifications

Quantity 1 g
Formula Weight 384.87
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrazolium Violet
RTECSNumber XF8080000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.