Learn More
Medchemexpress LLC Tetrindole mesylate | 170964-68-8 | 99.9% | 390.54 | C21H30N2O3S | 1 MG
SDP
Catalog No. 5000213388
Encompass_Preferred
Tetrindole mesylate is a research reagent that acts as a selective monoamine oxidase A (MAO A) inhibitor used in biochemical and pharmacological investigations of MAO-A activity and antidepressant mechanisms. It is supplied as a solid mesylate salt with high purity and is intended for laboratory research use only. Recommended storage: -20°C sealed and away from moisture; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Selective MAO-A inhibitory activity for mechanistic and pharmacology studies.
  • High purity (99.93%) suitable for analytical and preclinical assays.
  • Solid form for accurate weighing and formulation.
  • Contains CAS number 170964-68-8 for unambiguous identification.
  • Recommended cold storage to preserve stability.
Catalog No. 50-002-13388 Supplier Medchemexpress LLC Supplier No. HY1011691MG
Tetrindole mesylate is a research reagent that acts as a selective monoamine oxidase A (MAO A) inhibitor used in biochemical and pharmacological investigations of MAO-A activity and antidepressant mechanisms. It is supplied as a solid mesylate salt with high purity and is intended for laboratory research use only. Recommended storage: -20°C sealed and away from moisture; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Selective MAO-A inhibitory activity for mechanistic and pharmacology studies.
  • High purity (99.93%) suitable for analytical and preclinical assays.
  • Solid form for accurate weighing and formulation.
  • Contains CAS number 170964-68-8 for unambiguous identification.
  • Recommended cold storage to preserve stability.