Learn More
Bioworld TE (Tris-EDTA) Buffer, Sterile, 500 mL
Catalog No. 50196705
Encompass_Preferred
Product DescriptionTE (Tris-EDTA) is a sterile buffer that has a pH level of 8.0 brought up with HCl and is stored at room temperature. It contains 10 mM Tris and 1 mM EDTA. It is of biotechnology grade and no DNAse activity is detected.TE buffer is used to solubilize DNA or RNA while protecting it from degradation. Commonly used for DNA isolation and electrophoresis protocols.
Catalog No. 50-196-705 Supplier Bioworld Supplier No. 105300282
Add to Cart
Add to Cart
Product DescriptionTE (Tris-EDTA) is a sterile buffer that has a pH level of 8.0 brought up with HCl and is stored at room temperature. It contains 10 mM Tris and 1 mM EDTA. It is of biotechnology grade and no DNAse activity is detected.TE buffer is used to solubilize DNA or RNA while protecting it from degradation. Commonly used for DNA isolation and electrophoresis protocols.