Learn More
Cayman Chemical tetrnor-MIsoprostol 100ug
SDP
Catalog No. 502995420
Encompass_Preferred
A metabolite of misoprostol; formed when misoprostol undergoes rapid de-esterification to form misoprostol (free acid), which is further metabolized to form the more polar tetranor-misoprostol in rats, dogs, and monkeys
Catalog No. 50-299-5420 Supplier Cayman Chemical Supplier No. 22693100 UG
A metabolite of misoprostol; formed when misoprostol undergoes rapid de-esterification to form misoprostol (free acid), which is further metabolized to form the more polar tetranor-misoprostol in rats, dogs, and monkeys