Learn More
Cayman Chemical tetrnor-PGDM lacton-d6 100ug
SDP
Catalog No. 502951405
Encompass_Preferred
An internal standard for the quantification of tetranor-PGDM lactone by GC- or LC-MS
Catalog No. 50-295-1405 Supplier Cayman Chemical Supplier No. 11213100 UG
An internal standard for the quantification of tetranor-PGDM lactone by GC- or LC-MS