Learn More
Cayman Chemical tetrnor-PGEM 50ug
SDP
Catalog No. 502972894
Encompass_Preferred
A metabolite of PGE2; urine levels are increased in patients with diabetic nephropathy; increased urine levels are associated with a higher risk of breast cancer in postmenopausal women with a BMI of 2
Catalog No. 50-297-2894 Supplier Cayman Chemical Supplier No. 1484050 UG
A metabolite of PGE2; urine levels are increased in patients with diabetic nephropathy; increased urine levels are associated with a higher risk of breast cancer in postmenopausal women with a BMI of 2