Learn More
Cayman Chemical tetrnor-PGEM-d6 100ug
SDP
Catalog No. 502991753
Encompass_Preferred
An internal standard for the quantification of tetranor-PGEM by GC- or LC-MS
Catalog No. 50-299-1753 Supplier Cayman Chemical Supplier No. 314840100 UG
An internal standard for the quantification of tetranor-PGEM by GC- or LC-MS