Learn More
Cayman Chemical tetrnor-PGEM-d6 50ug
SDP
Catalog No. 502991676
Encompass_Preferred
An internal standard for the quantification of tetranor-PGEM by GC- or LC-MS
Catalog No. 50-299-1676 Supplier Cayman Chemical Supplier No. 31484050 UG
An internal standard for the quantification of tetranor-PGEM by GC- or LC-MS