Learn More
Cayman Chemical tetrnor-PGFM 100ug
SDP
Catalog No. 502967622
Encompass_Preferred
A major urinary metabolite of PGF2α
Catalog No. 50-296-7622 Supplier Cayman Chemical Supplier No. 16840100 UG
A major urinary metabolite of PGF2α