Learn More
Cayman Chemical tetrnorPGDM AChE Tracer 100dtn
SDP
Catalog No. 502942218
Encompass_Preferred
An IRF3 activator and adjuvant increases the mRNA expression of the genes encoding ISG54 and OASL in PH5CH8 cells and increases CXCL10 production in phorbol 12-myristate 13-acetate-differentiated THP-1 monocytes at 10 UM immunization with a vaccine in combination with KIN1148 increases blood IgG neutralizing and hemagglutinin antibody titers decreases PFUs in the lungs and lung-draining lymph nodes and increases the survival percentage of mice infected with a lethal dose of influenza virus when used as an adjuvant
Catalog No. 50-294-2218 Supplier Cayman Chemical Supplier No. 401000100 DTN
An IRF3 activator and adjuvant increases the mRNA expression of the genes encoding ISG54 and OASL in PH5CH8 cells and increases CXCL10 production in phorbol 12-myristate 13-acetate-differentiated THP-1 monocytes at 10 UM immunization with a vaccine in combination with KIN1148 increases blood IgG neutralizing and hemagglutinin antibody titers decreases PFUs in the lungs and lung-draining lymph nodes and increases the survival percentage of mice infected with a lethal dose of influenza virus when used as an adjuvant