Learn More
Cayman Chemical TetromycIn B 2.5mg
SDP
Catalog No. 502994261
Encompass_Preferred
A cysteine protease inhibitor (Kis = 0.62, 1.42, 32.5, and 1.59 μM for rhodesain, falcipain-2, cathepsin L, and cathepsin B, respectively); inhibits the growth of T. brucei in vitro (IC50 = 30.87 μM); cytotoxic to HEK293T kidney cells and J774.1 macrophages (IC50s = 71.77 and 20.2 μM, respectively)
Catalog No. 50-299-4261 Supplier Cayman Chemical Supplier No. 237662.5 MG
A cysteine protease inhibitor (Kis = 0.62, 1.42, 32.5, and 1.59 μM for rhodesain, falcipain-2, cathepsin L, and cathepsin B, respectively); inhibits the growth of T. brucei in vitro (IC50 = 30.87 μM); cytotoxic to HEK293T kidney cells and J774.1 macrophages (IC50s = 71.77 and 20.2 μM, respectively)