Learn More
Epigentek Inc tetX Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A50817100
Encompass_Preferred
HRP Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Clostridium tetani Tetanus toxin protein (123-573AA). Tested Applications: ELISA. Reactivity: Clostridium tetani. Alternative Names: Tentoxylysin, Tetanus toxin chain L, Tetanus toxin chain H, CTC_p60, tetX.
Catalog No. 50-119-7582