Learn More
AURORA BIOLABS LLC TEV Protease. 1000 units
Catalog No. NC3787967
Encompass
Tobacco Etch Virus nuclear-inclusion-a endopeptidase or TEV is highly sequence specific cysteine protease. It has a strict 7 amino acid cleavage recognition sequence of Glu-Asn-Leu-Tyr-Phe-Gln↓(Gly/Ser). The high specificity makes this protease excellent for the removal of affinity-tags from purified recombinant proteins. Purified TEV was expressed in E.coli and is shipped in 4°C packaging to preserve activity. TEV cleavage is best performed at room temperature in a P.H range of 4 – 9.
Catalog No. NC3787967 Supplier AURORA BIOLABS LLC Supplier No. 1900011K
Tobacco Etch Virus nuclear-inclusion-a endopeptidase or TEV is highly sequence specific cysteine protease. It has a strict 7 amino acid cleavage recognition sequence of Glu-Asn-Leu-Tyr-Phe-Gln↓(Gly/Ser). The high specificity makes this protease excellent for the removal of affinity-tags from purified recombinant proteins. Purified TEV was expressed in E.coli and is shipped in 4°C packaging to preserve activity. TEV cleavage is best performed at room temperature in a P.H range of 4 – 9.