Learn More
Genscript Corporation TEV Protease, His
Catalog No. 502272178
Encompass
Tobacco Etch Virus Protease is a highly site-specific cysteine protease that is found in the tags from fusion proteins. The optimal temperature for cleavage is 30°C; also it can be used at temperature as low as 4°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of recombinant viral TEV protease, reaction time, or incubation temperature. It can be removed by Ni2+ affinity resin. Recombinant Tobacco Etch Virus (TEV). The optimum recognition site for this enzyme is the sequence Glu-Asn-Leu-Tyr-Phe-Gln-(Gly/Ser) [ENLYFQ(G/S)] and cleavage occurs between the Gln and Gly/Ser residues, The most commonly used sequence is ENLYFQG. The protease is used to cleave affinity tags from fusion proteins. The optimal temperature for cleavage is 30°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of recombinant viral TEV protease, reaction time, or incubation temperature.
This item is not returnable. View return policy
Catalog No. 50-227-2178 Supplier Genscript Corporation Supplier No. Z030301K
Add to Cart
Add to Cart
Tobacco Etch Virus Protease is a highly site-specific cysteine protease that is found in the tags from fusion proteins. The optimal temperature for cleavage is 30°C; also it can be used at temperature as low as 4°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of recombinant viral TEV protease, reaction time, or incubation temperature. It can be removed by Ni2+ affinity resin. Recombinant Tobacco Etch Virus (TEV). The optimum recognition site for this enzyme is the sequence Glu-Asn-Leu-Tyr-Phe-Gln-(Gly/Ser) [ENLYFQ(G/S)] and cleavage occurs between the Gln and Gly/Ser residues, The most commonly used sequence is ENLYFQG. The protease is used to cleave affinity tags from fusion proteins. The optimal temperature for cleavage is 30°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of recombinant viral TEV protease, reaction time, or incubation temperature.