Learn More
ACROBiosystems TEV Protease (active enzyme, HPLC verified) 1000U
Catalog No. NC3738127
Encompass_Preferred
TEV Protease is a recombinant form of Tobacco Etch Virus protease expressed in Escherichia coli. This protease is used to cleave affinity tags from fusion proteins. The TEV Protease recognition sequence with the highest catalytic efficiency is ENLYFQS/G, the cleavage site is between Q and S/G. TEV protease contains N-terminus His-tag, it can be removed by Ni2+ affinity resin for purification of the target protein.
Catalog No. NC3738127 Supplier ACROBiosystems Supplier No. TEEV51431000U
TEV Protease is a recombinant form of Tobacco Etch Virus protease expressed in Escherichia coli. This protease is used to cleave affinity tags from fusion proteins. The TEV Protease recognition sequence with the highest catalytic efficiency is ENLYFQS/G, the cleavage site is between Q and S/G. TEV protease contains N-terminus His-tag, it can be removed by Ni2+ affinity resin for purification of the target protein.