Learn More
Eichrom Industries TEVA Resin, 50-100 µm, 2 mL, package of 50
SDP
Catalog No. NC1569049
Encompass
TEVA Resin has been applied on a routine basis to the analysis of technetium, the measurement of the tetravalent actinides, and the separation of americium from lanthanides. It is used alone or can be readily combined with other resins to perform more elaborate separations of multiple analytes.
Catalog No. NC1569049 Supplier Eichrom Industries Supplier No. TER50S
TEVA Resin has been applied on a routine basis to the analysis of technetium, the measurement of the tetravalent actinides, and the separation of americium from lanthanides. It is used alone or can be readily combined with other resins to perform more elaborate separations of multiple analytes.