Learn More
Aviva Systems Biology Corp TEX14 Antibody - C-terminal region (ARP53844_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53844_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEX14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TEX14. Alternate names: CT113ApplicationWB. Species reactivity: Horse, Human, Rabbit, Rat
Catalog No. 50-014-53734