Learn More
Aviva Systems Biology Corp TEX2 Antibody - N-terminal region (ARP49316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TEX2. Alternate names: DKFZp781G0721, HT008, KIAA1738, TMEM96ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53580