Learn More
Aviva Systems Biology Corp TEX264 Antibody - middle region (ARP47201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEX264. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TEX264. Alternate names: DKFZp451H0417, FLJ13935, SIG11, ZSIG11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59482