Learn More
Enterprise Technology Solutions Texas Instruments, Inc Financial Calculator, Advanced, w/ Case, 3"x6"x3/5", BK/GY
SDP
Catalog No. 5000740278
Encompass_Preferred
Easy-to-use Financial Calculator solves time-value-of-money calculations such as annuities, mortgages, leases, savings, and more. Generate amortization schedules. Compute NPV, IIR, break-even calculations, interest conversions, delta percent, profit and loss, cost, selling price, margin and markup. Performs cash-flow analysis for up to 24 uneven cash flows with up to four-digit frequencies. Calculator offers depreciation schedules; two day-count methods (actual or 30/360) to calculate bond price or yield to maturity or to call; list-based one-variable and two-variable statistics with four regression options: linear, logarithmic, exponential and power. Math functions include trigonometric calculations, natural logarithms, and powers. Design also features 10-digit display and 10 user memories. Automatic power-down conserves power. Calculator includes a protective slide case and one lithium 2032 battery.
Catalog No. 50-007-40278 Supplier Enterprise Technology Solutions Supplier No. TEXBAIIPLUS
Easy-to-use Financial Calculator solves time-value-of-money calculations such as annuities, mortgages, leases, savings, and more. Generate amortization schedules. Compute NPV, IIR, break-even calculations, interest conversions, delta percent, profit and loss, cost, selling price, margin and markup. Performs cash-flow analysis for up to 24 uneven cash flows with up to four-digit frequencies. Calculator offers depreciation schedules; two day-count methods (actual or 30/360) to calculate bond price or yield to maturity or to call; list-based one-variable and two-variable statistics with four regression options: linear, logarithmic, exponential and power. Math functions include trigonometric calculations, natural logarithms, and powers. Design also features 10-digit display and 10 user memories. Automatic power-down conserves power. Calculator includes a protective slide case and one lithium 2032 battery.