Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-15 Calculator
SDPSCI_ED

Catalog No. S65240
Encompass
Encompass_Preferred
Change view
Click to view available options
Display Type:
Digital
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
S65240 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65240 Supplier Texas Instruments Inc Supplier No. TI15
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Constant operations with counters help build tables.

  • Develops the concepts of multiplication, division, powers and units of measurement
  • Combines fraction capabilities with problem-solving function, place value and more
  • Two-line display shows entries and results at the same time
  • Review previous entries and results while allowing students to look for patterns

Order Info

Teacher pack also available (Cat. No. S90262TP).

Specifications

Product Type School Calculator
Display Type Digital
Color Black/Silver
Key Functions Powers and square root, Symbolic pi, convert pi to decimal, conversions between improper fractions and mixed numbers, conversions between fractions, decimals and percents
Material Plastic
Model TI-15
Power Supply Battery, Solar
For Use With (Application) Develops the concepts of multiplication, division, powers, and unit-of-measure concepts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.