Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-1706SV Calculator
SDPSCI_ED

Catalog No. S65245
Encompass
Encompass_Preferred
Change view
Click to view available options
Display Type:
Digital
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
S65245 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65245 Supplier Texas Instruments Inc Supplier No. TI1706
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economical, compact ANYLITE™ solar-powered memory calculator features four-function, slightly contoured keys for maximum speed and comfort.

  • Extra-large plus and minus keys
  • Operates even in dim light; never needs batteries
  • Attractive sliding case

 

Specifications

Product Type Basic Calculator
Display Type Digital
Keypad Color-coded, extra-large keys
Model TI-1706 SV
Dimensions (L x W) 4.8 x 2.7 in. (12.1 x 6.8cm)
Length (English) 4.8 in.
Length (Metric) 12.1cm
Width (English) 2.7 in.
Width (Metric) 6.8cm
Power Supply Solar powered with battery backup
For Use With (Application) General math, pre-algebra, algebra I and II, geometry, trigonometry, statistics, general science and biology
Includes Attractive sliding case
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.