Learn More
STAPLES ADVANTAGE Texas Instruments TI-30XIIS 10-Digit Scientific Battery & Solar Powered Scientific Calculator, Blue (TI30XIIS)
Catalog No. NC3806403
Encompass_Preferred
Scientific calculator two-line LCD display: 10-digit answer & two-digit exponent linePerfect choice for general math, statistics, pre-algebra, general scienceCalculator works from solar cell and one CR2025 battery
Catalog No. NC3806403 Supplier STAPLES ADVANTAGE Supplier No. 382647
Scientific calculator two-line LCD display: 10-digit answer & two-digit exponent linePerfect choice for general math, statistics, pre-algebra, general scienceCalculator works from solar cell and one CR2025 battery