Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-30XS MultiView™ Scientific Calculator
SDPSCI_ED

Catalog No. s96892
Encompass
Encompass_Preferred
Change view
Click to view available options
Display Type:
LCD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
S96892 LCD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96892 Supplier Texas Instruments Inc Supplier No. TI30XSMULTIBK
Only null left
Add to Cart
Add to Cart

Appropriate for general math, pre-algebra, algebra I and II, geometry, trigonometry, statistics, general science and biology.

Offers a four-line display to view multiple calculations and answers simultaneously. Easily scroll, review and edit current or previous entries.

  • MultiView™ display
  • Toggle key
  • Scrolling and editing
  • Stacked fractions
  • Standard mathematical notation
  • Scientific notation input and output
  • Vertical menus
  • Data/list editor

Specifications

Product Type Scientific Calculator
Display Type LCD
Grade High School
Keypad Color-coded, hard plastic keys
Model TI-30XS
Power Supply Solar powered with battery backup
For Use With (Application) For high school math and science exploration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.