Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-34 Calculator
SDPSCI_ED

Catalog No. S97827
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Scientific Calculator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S97827 Scientific Calculator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S97827 Supplier Texas Instruments Inc Supplier No. TI34MVBK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Two-line display with fraction capability helps with exploring math and science concepts.

  • Top line displays up to 11 characters
  • Bottom line displays up to 10 digits with two-digit exponent
Key features:
  • Previous entry review
  • Advanced menus
  • Fraction features
  • Two constant keys with counter
  • Basic scientific and trigonometric functions

Specifications

Product Type Scientific Calculator
Display Type LCD
Grade Middle School
Keypad Color-coded, hard plastic keys
Model TI-34
Power Supply Solar
For Use With (Application) For use in middle grades math and science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.