Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-503SV Calculator
SDPSCI_ED

Catalog No. S65247
Encompass
Encompass_Preferred
Change view
Click to view available options
Display Type:
Digital
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
S65247 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65247 Supplier Texas Instruments Inc Supplier No. TI503PLUS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • SuperView™ display provides easy, on-the-spot answers
  • Roomy keyboard with well-spaced rubber keys
  • Sign key (+/-) simplifies entry of negative numbers
  • Automatic power-down feature conserves battery life

Order Info

Battery-powered, pocket-size calculator is perfect for general-purpose use.

Specifications

Product Type Basic Calculator
Display Type Digital
Keypad Roomy keyboard with well-spaced rubber keys
Model TI-503SV
Dimensions (L x W) 4.5 x 2.8 in. (11.4 x 7.1cm)
Length (English) 4.5 in.
Length (Metric) 11.4cm
Width (English) 2.8 in.
Width (Metric) 7.1cm
Power Supply D389 Battery
For Use With (Application) General-purpose use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.