Learn More
Abcam Te x as Red Conjugation Kit (Fast) - Lightning-Link, 3 x 100UG
Catalog No. 502589600
Encompass_Preferred
Texas Red conjugation / labeling in < 20 mins with 30 secs hands-on time using Texas Red Conjugation Kit (Fast) - Lightning-Link ab195225. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9600 Supplier Abcam Supplier No. AB1952253X100UG
Texas Red conjugation / labeling in < 20 mins with 30 secs hands-on time using Texas Red Conjugation Kit (Fast) - Lightning-Link ab195225. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement