Learn More
Biotium Texas Red-DHPE
Catalog No. 501964722
Encompass_Preferred
Texas Red-DHPE (N-(Texas Red sulfonyl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt) is a rhodamine-labeled glycerophosphoethanolamine lipid. DHPE conjugates of rhodamine dyes have been used as fluorescence energy acceptors in combination with NBDPE as the donor in membrane fusion assays using the principle of fluorescence energy transfer (FRET). These probes have also been used for following membrane trafficking during endocytosis.Properties: Ex/Em(MeOH) = 582/601 nm; Dark solid soluble in chloroform; Store at -20°C and protect from light; C74H117N4O14PS2; MW: 1380.77; [187099-99-6].Texas Red is a registerd trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4722 Supplier Biotium Supplier No. 60027
Texas Red-DHPE (N-(Texas Red sulfonyl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt) is a rhodamine-labeled glycerophosphoethanolamine lipid. DHPE conjugates of rhodamine dyes have been used as fluorescence energy acceptors in combination with NBDPE as the donor in membrane fusion assays using the principle of fluorescence energy transfer (FRET). These probes have also been used for following membrane trafficking during endocytosis.Properties: Ex/Em(MeOH) = 582/601 nm; Dark solid soluble in chloroform; Store at -20°C and protect from light; C74H117N4O14PS2; MW: 1380.77; [187099-99-6].Texas Red is a registerd trademark of Thermo Fisher Scientific.