Learn More
Vector Laboratories Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Texas Red 1 mg
SDP
Catalog No. NC9921836
Encompass
Lycopersicon esculentum lectin (LEL or tomato lectin, TL), binds to glycophorin and Tamm-Horsfall glycoprotein and has been used effectively to label and image vascular endothelium in rodents. This Texas Red conjugate features a ratio of fluorochrome to lectin protein that provides optimal staining (excitation 595 nm, emission 615 nm). It is supplied essentially free of unconjugated biotins and preserved with sodium azide.
Catalog No. NC9921836 Supplier Vector Laboratories Supplier No. TL11761
Lycopersicon esculentum lectin (LEL or tomato lectin, TL), binds to glycophorin and Tamm-Horsfall glycoprotein and has been used effectively to label and image vascular endothelium in rodents. This Texas Red conjugate features a ratio of fluorochrome to lectin protein that provides optimal staining (excitation 595 nm, emission 615 nm). It is supplied essentially free of unconjugated biotins and preserved with sodium azide.