Learn More
Aat Bioquest Texas Red® cadaverine *Single Isomer*
SDP
Catalog No. 501952900
Encompass_Preferred
Texas Red® cadaverine reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates. It is widely used for labeling glycoproteins and carbohydrates through reductive amination.
Catalog No. 50-195-2900 Supplier Aat Bioquest Supplier No. 482
Texas Red® cadaverine reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates. It is widely used for labeling glycoproteins and carbohydrates through reductive amination.