Learn More
Aat Bioquest Texas Red-streptavidin conjugate (1 mg)
SDP
Catalog No. 502249620
Encompass_Preferred
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins protein motifs nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9620 Supplier Aat Bioquest Supplier No. 16911
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins protein motifs nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin. This catalog is for 1 mg.