Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Texas Red™ Sulfonyl Chloride, mixed isomers

Catalog No. p-7076296
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1905 10 x 1 mg
T353 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1905 Supplier Invitrogen™ Supplier No. T1905
Only null left
Add to Cart
Edge
Add to Cart

Texas Red™ Sulfonyl Chloride, mixed isomers

The amine-reactive Texas Red™ sulfonyl chloride can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 1 mg
Product Type Sulfonyl Chloride
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Sulfonyl Chloride
Label or Dye Texas Red™
Label Type Classic Dyes
Product Line Texas Red
Emission 615
Excitation 595
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.