Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Texas Red™-X, Succinimidyl Ester, mixed isomers

Catalog No. T6134
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T6134 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T6134 Supplier Invitrogen™ Supplier No. T6134
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Texas Red™-X, Succinimidyl Ester, mixed isomers

The isomeric mixture of amine-reactive Texas Red-X, succinimidyl ester and its conjugates emit at a longer wavelength than do either tetramethylrhodamine or Lissamine rhodamine B, and are among the most commonly used long-wavelength third labels in fluorescence microscopy and flow cytometry. Protein conjugates prepared with Texas Red-X succinimidyl ester also have a higher fluorescence yield than those with the same labeling ratio prepared with Texas Red sulfonyl chloride.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Texas Red™
Label Type Classic Dyes
Product Line Texas Red

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.