Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Texas Red™-X, Succinimidyl Ester, single isomer

Catalog No. T20175
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20175 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20175 Supplier Invitrogen™ Supplier No. T20175
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Texas Red™-X, Succinimidyl Ester, single isomer

The amine-reactive Texas Red™-X, succinimidyl ester can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm. This reactive dye contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Texas Red™
Label Type Classic Dyes
Product Line Texas Red
Emission 615
Excitation 595
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.