Learn More
Texwipe™ TexQ™ Quaternary Ammonium Disinfectants
SDP
Catalog No. 19034970 Shop All Texwipe Products
Change view
Click to view available options
Container Type:
Bottle
Catalog No. Container Type
19-034-970 Bottle
1 options
This item is not returnable. View return policy
Catalog No. 19-034-970 Supplier Texwipe™ Supplier No. TX650
Add to Cart
Add to Cart
This item is not returnable. View return policy
One step cleaner and disinfectant
  • Dye/fragrance free
  • Effective against bacteria, virus and fungi
  • Available in a 22 oz. bottle or 1 gallon concentrate
  • EPA registered
Specifications
Sterility Sterile
Certifications/Compliance EPA
Container Type Bottle
For Use With (Application) Effective against a broad spectrum of Bacteria, Virus and Fungi and inhibits the growth of mold and mildew and their odors
Volume (English) 22 oz.