Learn More
Texwipe™ TexTab™ Disinfectant Tablets
SDP
Catalog No. 18368141 Shop All Texwipe Products
Change view
Click to view available options
Packaging Quantity:
256 Tablets/Bottle
Catalog No. Packaging Quantity
18-368-141 256 Tablets/Bottle
1 options
This item is not returnable. View return policy
Catalog No. 18-368-141 Supplier Texwipe™ Supplier No. TX6460
This item is not returnable. View return policy
Broad spectrum and sporicidal disinfectant in an effervescent tablet form.
  • Sporocide
  • Kills Clostridium difficile (C. diff) spores in four minutes
  • Effervescent tablet form
  • Exact dosage tablet to deliver an accurate strength solution
  • Eliminates risk of concentrated bleach spills
  • Sporicidal strength: weekly/biweekly use
  • Safe for users
  • Does not damage finishes or equipment
  • Pathogens eliminated: C. difficale spores, mycobacteria, Gram-negative and Gram-positive bacteria, antibiotic-resistant bacteria
Applications:
  • Disinfecting any hard, non-porous, inanimate surface
  • Replacing liquid bleach solutions in all applications
  • Using as part of a disinfectant rotation program
  • Pre-cleaning gross soil before disinfection
  • Cleaning and disinfecting small work areas as well as
    large areas (floors, walls, ceilings)
  • Fogging
  • Used in pharmaceutical, medical device, cosmetic manufacturing facilities
Specifications
Product Type Disinfectant Tablets
For Use With (Application) Kills spores and germs
Packaging Quantity 256 Tablets/Bottle