Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ TexTra™ Non-sterile Dry Cleanroom Wipers
SDP

Catalog No. 17000336 Shop All Texwipe Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
17-000-336 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-000-336 Supplier Texwipe™ Supplier No. TX2424
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dry cleanroom wipers with high absorbency for spill control, rigorous wiping and solution application.

TexTra™ Non-sterile Dry Cleanroom Wipers are made from 100% polyester with a cut edge. These wipers are made from thick, cleanroom polyester knit. These single-ply wipers have the liquid-holding capacity of two-ply wipers. TexTra™ combines this high absorbency with great abrasion resistance, resulting in a wiper that is great for wiping and cleaning surfaces, equipment, and parts. Furthermore, it is suitable in applying and removing lubricants, adhesives, residues, and other solutions including disinfectants.

  • High absorbency wiper ideal for spill control, cleaning, and solution application
  • Cleanroom laundered for low levels of ions and NVRs (non-volatile residues) particles
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Meets USP 797 and USP 800 wiper requirements

Specifications

Material Polyester
Autoclavable Autoclavable
Certifications/Compliance ISO Class 3 to 8, Class 1 to 100,000
Color White
Edges Cut
For Use With (Application) Cleaning and Solution Application
Sterility Non-sterile
Certifications/Compliance (Europe) Grade A to D
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.