Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ AlphaMop™ Replacement Handle
SDP

Catalog No. 19222373 Shop All Texwipe Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-222-373 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-222-373 Supplier Texwipe™ Supplier No. TX7123
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Low profile, swivel head with lightweight handles enable easier cleaning of floors, walls, ceilings, and hard-to-reach areas

  • 60 in. (1.5 m)
  • Fiberglass
  • White
  • Specifications

    Product Type Replacement Handle
    Color White
    Sterility Non-sterile
    Autoclavable Autoclavable
    For Use With (Equipment) AlphaMop
    Color-coded No
    Material Fiberglass
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.