Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ AlphaMop™ Replacement Heads
SDP

Catalog No. 19222366 Shop All Texwipe Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Not Autoclavable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-222-366 Autoclavable
19-222-367 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-366 Supplier Texwipe™ Supplier No. TX7108AH
Add to Cart
Add to Cart

AlphaMop™ 15 x 8 in. (38 x 20 cm) Thermoplastic Mop Head

  • The large mop head allows for wider surface coverage and faster cleaning results
  • Quick-change system allows the user to replace covers / pads frequently, adhering to strict cleaning protocols
  • The polyester pad assures the mop cover to conform to surfaces for consistent cleaning
  • Specifications

    Product Type Replacement Head
    Sterility Non-sterile
    Autoclavable Autoclavable
    For Use With (Equipment) AlphaMop
    Color-coded No
    Material Thermoplastic
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.