Learn More
Texwipe™ AlphaMop™ Replacement Microdenier Mop Covers
SDP
Catalog No. 19139876 Shop All Texwipe Products
Change view
Click to view available options
Material:
Microdenier Polyester Fiber
Catalog No. Material
19-139-876 Microdenier Polyester Fiber
1 options
Catalog No. 19-139-876 Supplier Texwipe™ Supplier No. TX7118M
Specifications
Product Type Mop Cover
Sterility Non-sterile
Autoclavable Autoclavable
Includes Foam Pads
For Use With (Application) Maximum removal of small and micro contaminants and used in cleanrooms, disinfecting with quat-based cleaners, pharmaceuticals, biologics, medical device
Material Microdenier Polyester Fiber