Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ Isolator Cleaning Tool Replacement Telescoping Handle
SDP

Catalog No. 19222372 Shop All Texwipe Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-222-372 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-222-372 Supplier Texwipe™ Supplier No. TX7122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Replacement Telescoping Handle
Color White
Autoclavable Autoclavable
For Use With (Equipment) Isolator Cleaning Tool
Material Fiberglass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.