Learn More
Advantus Texwipe™ Tape Head Wipes
SDP
Catalog No. 18389 Shop All Advantus Corporation Products
Click to view available options
For Use With (Equipment):
Tape transport systems
Catalog No. For Use With (Equipment)
18-389 Tape transport systems
1 options
Catalog No. 18-389 Supplier Advantus Supplier No. CT801
Safely remove oxide buildup and particulates from tape transport systems
  • Pre-saturated TexPad™ wipe (91% isopropyl alcohol and 9% deionized water) is non-abrasive and lint free
  • Evaporates quickly, so equipment is ready to use
  • Premeasured, single-use wipe assures no cross-contamination of equipment
  • 100% polyester wipe
  • 80 individually wrapped wipes per box
Specifications
Product Type Tape Head Cleaner
Product Line TexPad
Disposable Single-use
For Use With (Application) Removal of dust, dirt and Magnetic opxide buildup from tape heads, guides, capstans and magnetic disc surfaces, cleaning of optic fiber connectors
For Use With (Equipment) Tape transport systems
Material Polyester