Learn More
Texwipe™ TexWrite™ Clipboard
SDP
Catalog No. 191202807 Shop All Texwipe Products
Change view
Click to view available options
Material:
Polypropylene, Plastic
Catalog No. Material
19-120-2807 Polypropylene, Plastic
1 options
This item is not returnable. View return policy
Catalog No. 19-120-2807 Supplier Texwipe™ Supplier No. TX5835
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal portable writing surface for use in critical environments
  • Corrosion- and chemical-resistant, durable polypropylene construction
  • No sharp edges to damage gloves or garments
  • Rigid back allows use while standing
  • Suitable for use with 8 1/2 × 11 in. paper
  • Securely holds up to 50 sheets of cleanroom paper
  • Cleanroom packaged
Specifications
Material Polypropylene, Plastic
For Use With (Application) 8.5 x 11 in. or A4 paper
Product Type Cleanroom Clipboard