Learn More
Description
- Produced, packed in ISO Class 4 cleanroom
- Hot cut
- Thermally sealed edges
- Continuous-filament
- Double-knit
- Diamensions: 23 × 23cm (9 × 9 in.)
Specifications
Specifications
| Product Type | Thermaseal 70 Wipers |
| Length (English) | 9 in. |
| Width (English) | 9 in. |
| Material | Polyester |
| Dimensions (L x W) | 9 x 9 in. |
| Packaging Quantity | 150 Wipers/Bag |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
