Learn More
Andwin Scientific TEXWIPES 150/PK
SDP
Catalog No. 501305854
Encompass_Preferred
The 12x12 Texwipe low-particulate wipes is used for wiping aqueous spills and cleaning precision laboratory components. It is constructed from nonwoven cellulose and polyester blend, allowing excellent solvent compatibility. It is sold for a pack of 150.
This item is not returnable. View return policy
Catalog No. 50-130-5854 Supplier Andwin Scientific Supplier No. 3367702A
The 12x12 Texwipe low-particulate wipes is used for wiping aqueous spills and cleaning precision laboratory components. It is constructed from nonwoven cellulose and polyester blend, allowing excellent solvent compatibility. It is sold for a pack of 150.