Learn More
Cole-Parmer Texwipe TX612 Low-particulate wipes, 12" x 12", 150/bag
Catalog No. NC9857092 Shop All Cole-Parmer Products
Encompass_Preferred
Low-particulate TechniCloth is more durable, absorbent, and softer than paper wipes. 12" x 12" wipe size; 150 wipes per bag. ISO Class 5 compliant.
This item is not returnable. View return policy
Catalog No. NC9857092 Supplier Cole-Parmer Supplier No. 3367702
Low-particulate TechniCloth is more durable, absorbent, and softer than paper wipes. 12" x 12" wipe size; 150 wipes per bag. ISO Class 5 compliant.