Learn More
Medchemexpress LLC HY-15448 10mg Medchemexpress, Tezacaftor CAS:1152311-62-0 Purity:>98%
SDP
Catalog No. 502029125
Encompass_Preferred
Medchemexpress, HY-15448 10mg Tezacaftor CAS:1152311-62-0 Tezacaftor (VX-661) is a second F508del CFTR corrector and help CFTR protein reach the cell surface. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9125 Supplier Medchemexpress LLC Supplier No. HY1544810MG
Medchemexpress, HY-15448 10mg Tezacaftor CAS:1152311-62-0 Tezacaftor (VX-661) is a second F508del CFTR corrector and help CFTR protein reach the cell surface. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.